siRNA Details
| Gene Symbol: Agk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127497.1 | NP_001120969.1 | 14 | 1068 | ||
| XM_006236363.2 | XP_006236425.1 | 14 | 1370 | |||
| XM_008762821.1 | XP_008761043.1 | 14 | 1465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agk| Gene Symbol: Agk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127497.1 | NP_001120969.1 | 14 | 1068 | ||
| XM_006236363.2 | XP_006236425.1 | 14 | 1370 | |||
| XM_008762821.1 | XP_008761043.1 | 14 | 1465 | |||