siRNA Details
| Gene Symbol: Tas2r143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025061.1 | NP_001020232.1 | 1 | 838 | ||
| Gene Symbol: LOC103690198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762927.1 | XP_008761149.1 | 1 | 1166 | ||

Tas2r143
Genomic Map