siRNA Details
| Gene Symbol: Zfp777 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109348.1 | NP_001102818.1 | 5 | 1705 | ||
| XM_006236443.2 | XP_006236505.1 | 6 | 1945 | |||
| XM_008762913.1 | XP_008761135.1 | 6 | 1734 | |||
| XM_008762914.1 | XP_008761136.1 | 6 | 1759 | |||

Zfp777
Genomic Map