siRNA Details
| Gene Symbol: Cpvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029927.1 | NP_001025098.1 | 4 | 414 | ||
| XM_006236511.2 | XP_006236573.2 | 4 | 890 | |||
| XM_008762915.1 | XP_008761137.1 | 6 | 1199 | |||
| GenBank mRNA | BC097471.1 | - | 4 | 414 | 7457768 | |

Cpvl
Genomic Map