siRNA Details
| Gene Symbol: Gp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031825.1 | NP_001026995.1 | 2 | 221 | ||
| GenBank mRNA | AB158423.1 | - | 2 | 221 | ||
| AB158424.1 | - | 2 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gp9| Gene Symbol: Gp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031825.1 | NP_001026995.1 | 2 | 221 | ||
| GenBank mRNA | AB158423.1 | - | 2 | 221 | ||
| AB158424.1 | - | 2 | 221 | |||