siRNA Details
| Gene Symbol: Jagn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044272.1 | NP_001037737.1 | 2 | 326 | ||
| XM_006237053.2 | XP_006237115.1 | 2 | 375 | |||
| GenBank mRNA | BC098749.1 | - | 2 | 326 | 7365215 | |
| FQ225993.1 | - | 2 | 330 | |||

Jagn1
Genomic Map