siRNA Details
| Gene Symbol: Jagn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044272.1 | NP_001037737.1 | 2 | 595 | ||
| GenBank mRNA | BC098749.1 | - | 2 | 595 | 7365215 | |
| FQ225993.1 | - | 2 | 599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jagn1| Gene Symbol: Jagn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044272.1 | NP_001037737.1 | 2 | 595 | ||
| GenBank mRNA | BC098749.1 | - | 2 | 595 | 7365215 | |
| FQ225993.1 | - | 2 | 599 | |||