siRNA Details
| Gene Symbol: Prrt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749809.3 | XP_003749857.1 | 2 | 753 | ||
| XM_006237062.2 | XP_006237124.1 | 2 | 831 | |||
| XM_006237064.2 | XP_006237126.1 | 2 | 676 | |||
| XM_008763200.1 | XP_008761422.1 | 1 | 675 | |||

Prrt3
Genomic Map