siRNA Details
| Gene Symbol: Foxj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109352.1 | NP_001102822.1 | 8, 9 | 2134 | ||
| XM_006237311.2 | XP_006237373.1 | 9, 10 | 1947 | |||
| GenBank mRNA | FQ212196.1 | - | 1, 2 | 382 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxj2| Gene Symbol: Foxj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109352.1 | NP_001102822.1 | 8, 9 | 2134 | ||
| XM_006237311.2 | XP_006237373.1 | 9, 10 | 1947 | |||
| GenBank mRNA | FQ212196.1 | - | 1, 2 | 382 | ||