siRNA Details
| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109354.1 | NP_001102824.1 | 4 | 548 | ||
| GenBank mRNA | GU357486.1 | - | 4 | 452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec9a| Gene Symbol: Clec9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109354.1 | NP_001102824.1 | 4 | 548 | ||
| GenBank mRNA | GU357486.1 | - | 4 | 452 | ||