siRNA Details
Gene Symbol: Clec9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109354.1 | NP_001102824.1 | 1, 2 | 179 | ||
XM_006237099.2 | XP_006237161.1 | 1, 2 | 1055 | |||
XM_008763326.1 | XP_008761548.1 | 1, 2 | 56 | |||
GenBank mRNA | GU357486.1 | - | 1, 2 | 83 |
