siRNA Details
| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173386.1 | NP_001166857.1 | 2, 3 | 251 | ||
| GenBank mRNA | AB455106.1 | - | 2, 3 | 343 | ||
| BC166774.1 | - | 2, 3 | 251 | |||
| GU357485.1 | - | 2, 3 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec7a| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173386.1 | NP_001166857.1 | 2, 3 | 251 | ||
| GenBank mRNA | AB455106.1 | - | 2, 3 | 343 | ||
| BC166774.1 | - | 2, 3 | 251 | |||
| GU357485.1 | - | 2, 3 | 183 | |||