siRNA Details
| Gene Symbol: LOC690097 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763433.1 | XP_008761655.1 | 8 | 1386 | ||
| Gene Symbol: LOC497796 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111310.1 | NP_001104780.1 | 1119 | |||
| GenBank mRNA | BC093399.1 | - | 1119 | 7389533 | ||

LOC690097
Genomic Map