siRNA Details
Gene Symbol: LOC502908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763439.1 | XP_008761661.1 | 4 | 926 | ||
GenBank mRNA | AY669088.1 | - | 4 | 464 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC502908 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763439.1 | XP_008761661.1 | 4 | 926 | ||
GenBank mRNA | AY669088.1 | - | 4 | 464 |