siRNA Details
| Gene Symbol: LOC502908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763439.1 | XP_008761661.1 | 5 | 1025 | ||
| GenBank mRNA | AY669088.1 | - | 5 | 563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC502908| Gene Symbol: LOC502908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763439.1 | XP_008761661.1 | 5 | 1025 | ||
| GenBank mRNA | AY669088.1 | - | 5 | 563 | ||