siRNA Details
| Gene Symbol: Arid3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173981.1 | NP_001167452.1 | 3, 4 | 567 | ||
| XM_008763641.1 | XP_008761863.1 | 4, 5 | 781 | |||
| XM_008763642.1 | XP_008761864.1 | 4, 5 | 781 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arid3c| Gene Symbol: Arid3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173981.1 | NP_001167452.1 | 3, 4 | 567 | ||
| XM_008763641.1 | XP_008761863.1 | 4, 5 | 781 | |||
| XM_008763642.1 | XP_008761864.1 | 4, 5 | 781 | |||