siRNA Details
| Gene Symbol: LOC100911527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749962.1 | XP_003750010.1 | 1 | 509 | ||
| Gene Symbol: RGD1560539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749961.3 | XP_003750009.1 | 1 | 592 | ||

LOC100911527
Genomic Map