siRNA Details
| Gene Symbol: Ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173388.1 | NP_001166859.1 | 5 | 983 | ||
| XM_006239436.2 | XP_006239498.1 | 4 | 970 | |||
| GenBank mRNA | BC098773.1 | - | 4 | 833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnnbip1| Gene Symbol: Ctnnbip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173388.1 | NP_001166859.1 | 5 | 983 | ||
| XM_006239436.2 | XP_006239498.1 | 4 | 970 | |||
| GenBank mRNA | BC098773.1 | - | 4 | 833 | ||