siRNA Details
| Gene Symbol: Arfgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044273.1 | NP_001037738.1 | 11 | 1107 | ||
| GenBank mRNA | BC099081.1 | - | 11 | 1107 | 7443703 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arfgap3| Gene Symbol: Arfgap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044273.1 | NP_001037738.1 | 11 | 1107 | ||
| GenBank mRNA | BC099081.1 | - | 11 | 1107 | 7443703 | |