siRNA Details
| Gene Symbol: LOC102549491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257401.2 | XP_006257463.1 | 1 | 2205 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102549491| Gene Symbol: LOC102549491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257401.2 | XP_006257463.1 | 1 | 2205 | ||