siRNA Details
| Gene Symbol: LOC100362820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765885.1 | XP_008764107.1 | 4 | 3553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100362820| Gene Symbol: LOC100362820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765885.1 | XP_008764107.1 | 4 | 3553 | ||