siRNA Details
| Gene Symbol: Iqcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109361.1 | NP_001102831.1 | 3 | 294 | ||
| XM_006243838.2 | XP_006243900.1 | 2 | 1371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iqcf1| Gene Symbol: Iqcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109361.1 | NP_001102831.1 | 3 | 294 | ||
| XM_006243838.2 | XP_006243900.1 | 2 | 1371 | |||