siRNA Details
Gene Symbol: Rab17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109363.2 | NP_001102833.1 | 3, 4 | 487 | ||
GenBank mRNA | BC167022.1 | - | 3, 4 | 487 | 7126868 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109363.2 | NP_001102833.1 | 3, 4 | 487 | ||
GenBank mRNA | BC167022.1 | - | 3, 4 | 487 | 7126868 |