siRNA Details
| Gene Symbol: LOC100364769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177828.1 | NP_001171299.1 | 10 | 1067 | ||
| GenBank mRNA | AY539887.1 | - | 10 | 1067 | ||
| Gene Symbol: LOC257642 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY539885.1 | - | 191 | |||

LOC100364769
Genomic Map