siRNA Details
| Gene Symbol: RGD1563528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109365.1 | NP_001102835.1 | 1 | 418 | ||
| XM_006256674.2 | XP_006256736.1 | 1 | 3837 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563528| Gene Symbol: RGD1563528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109365.1 | NP_001102835.1 | 1 | 418 | ||
| XM_006256674.2 | XP_006256736.1 | 1 | 3837 | |||