siRNA Details
| Gene Symbol: LOC503435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109366.1 | NP_001102836.1 | 1 | 500 | ||
| XM_006256820.2 | XP_006256882.1 | 2 | 1191 | |||
| XM_008773133.1 | XP_008771355.1 | 2 | 737 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC503435| Gene Symbol: LOC503435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109366.1 | NP_001102836.1 | 1 | 500 | ||
| XM_006256820.2 | XP_006256882.1 | 2 | 1191 | |||
| XM_008773133.1 | XP_008771355.1 | 2 | 737 | |||