siRNA Details
| Gene Symbol: Mageb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109367.1 | NP_001102837.1 | 1 | 183 | ||
| XM_006256984.2 | XP_006257046.1 | 2 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mageb7| Gene Symbol: Mageb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109367.1 | NP_001102837.1 | 1 | 183 | ||
| XM_006256984.2 | XP_006257046.1 | 2 | 399 | |||