siRNA Details
| Gene Symbol: LOC100910698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752110.3 | XP_003752158.1 | 7 | 1115 | ||
| XM_008773459.1 | XP_008771681.1 | 8 | 1043 | |||
| Gene Symbol: Pramel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109368.1 | NP_001102838.1 | 2 | 604 | ||

LOC100910698
Genomic Map