siRNA Details
Gene Symbol: Cd300lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167664.1 | NP_001161136.1 | 4 | 563 | ||
GenBank mRNA | GU054494.1 | - | 4 | 563 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd300lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167664.1 | NP_001161136.1 | 4 | 563 | ||
GenBank mRNA | GU054494.1 | - | 4 | 563 |