siRNA Details
| Gene Symbol: Klri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012648.1 | NP_001012666.1 | 5 | 600 | ||
| XM_008763327.1 | XP_008761549.1 | 4 | 844 | |||
| GenBank mRNA | AY324871.1 | - | 5 | 600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klri2| Gene Symbol: Klri2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012648.1 | NP_001012666.1 | 5 | 600 | ||
| XM_008763327.1 | XP_008761549.1 | 4 | 844 | |||
| GenBank mRNA | AY324871.1 | - | 5 | 600 | ||