siRNA Details
| Gene Symbol: Klri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012649.1 | NP_001012667.1 | 2 | 278 | ||
| XM_008763328.1 | XP_008761550.1 | 2 | 347 | |||
| GenBank mRNA | AY286494.1 | - | 2 | 278 | ||
| FQ233804.1 | - | 2 | 296 | |||

Klri1
Genomic Map