siRNA Details
| Gene Symbol: Dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001053826.4 | XP_001053826.1 | 3 | 710 | ||
| XM_006242286.2 | XP_006242348.1 | 2 | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbx2| Gene Symbol: Dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001053826.4 | XP_001053826.1 | 3 | 710 | ||
| XM_006242286.2 | XP_006242348.1 | 2 | 614 | |||