siRNA Details
| Gene Symbol: Ncald | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024371.1 | NP_001019542.1 | 2 | 405 | ||
| XM_008765456.1 | XP_008763678.1 | 3 | 395 | |||
| GenBank mRNA | BC087105.1 | - | 2 | 405 | 7193019 | |
| FQ214436.1 | - | 414 | ||||

Ncald
Genomic Map