siRNA Details
| Gene Symbol: Akirin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030054.1 | NP_001025225.1 | 3, 4 | 603 | ||
| GenBank mRNA | CB761626.1 | - | 1, 2 | 157 | ||
| CV107792.1 | - | 3, 4 | 603 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akirin1| Gene Symbol: Akirin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030054.1 | NP_001025225.1 | 3, 4 | 603 | ||
| GenBank mRNA | CB761626.1 | - | 1, 2 | 157 | ||
| CV107792.1 | - | 3, 4 | 603 | |||