siRNA Details
| Gene Symbol: LOC606294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031627.1 | NP_001026797.1 | 2 | 465 | ||
| GenBank mRNA | BC060521.1 | - | 2 | 474 | ||
| BC098692.1 | - | 2 | 465 | 7386167 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC606294| Gene Symbol: LOC606294 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031627.1 | NP_001026797.1 | 2 | 465 | ||
| GenBank mRNA | BC060521.1 | - | 2 | 474 | ||
| BC098692.1 | - | 2 | 465 | 7386167 | ||