siRNA Details
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033079.1 | NP_001028251.1 | 11 | 2178 | ||
GenBank mRNA | L06434.2 | - | 11 | 2178 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc6a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033079.1 | NP_001028251.1 | 11 | 2178 | ||
GenBank mRNA | L06434.2 | - | 11 | 2178 |