siRNA Details
| Gene Symbol: Med24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034079.1 | NP_001029251.1 | 22, 23 | 2605 | ||
| XM_006247415.2 | XP_006247477.1 | 23, 24 | 2702 | |||
| XM_006247416.2 | XP_006247478.1 | 23, 24 | 2748 | |||
| GenBank mRNA | BC097461.1 | - | 22, 23 | 2605 | 7457533 | |
| FQ210692.1 | - | 22, 23 | 2505 | |||

Med24
Genomic Map