siRNA Details
Gene Symbol: Ppapdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034854.1 | NP_001030026.1 | 1 | 324 | ||
GenBank mRNA | BC081970.1 | - | 1 | 324 | 7125531 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppapdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034854.1 | NP_001030026.1 | 1 | 324 | ||
GenBank mRNA | BC081970.1 | - | 1 | 324 | 7125531 |