siRNA Details
| Gene Symbol: Gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034855.1 | NP_001030027.1 | 6 | 2624 | ||
| XM_006239485.2 | XP_006239547.1 | 6 | 5029 | |||
| XM_006239486.2 | XP_006239548.1 | 6 | 4960 | |||
| GenBank mRNA | BC105872.1 | - | 6 | 2624 | 7133996 | |

Gpr153
Genomic Map