siRNA Details
| Gene Symbol: Acsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034951.1 | NP_001030123.1 | 14, 15 | 1866 | ||
| GenBank mRNA | BC099826.1 | - | 14, 15 | 1866 | 7934093 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acsf2| Gene Symbol: Acsf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034951.1 | NP_001030123.1 | 14, 15 | 1866 | ||
| GenBank mRNA | BC099826.1 | - | 14, 15 | 1866 | 7934093 | |