siRNA Details
Gene Symbol: Lrrc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037336.1 | NP_001032413.1 | 1 | 2009 | ||
GenBank mRNA | DQ119102.1 | - | 1 | 2009 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138682, AK046781, AF300458, DQ177325, BC117833, and others.
Gene Ontology Categories:
