siRNA Details
| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044274.1 | NP_001037739.1 | 3, 4 | 152 | ||
| GenBank mRNA | DQ198380.1 | - | 3, 4 | 152 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odam| Gene Symbol: Odam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044274.1 | NP_001037739.1 | 3, 4 | 152 | ||
| GenBank mRNA | DQ198380.1 | - | 3, 4 | 152 | ||