siRNA Details
| Gene Symbol: Defb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037527.1 | NP_001032616.1 | 2 | 157 | ||
| GenBank mRNA | AY621372.1 | - | 2 | 157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Defb49| Gene Symbol: Defb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037527.1 | NP_001032616.1 | 2 | 157 | ||
| GenBank mRNA | AY621372.1 | - | 2 | 157 | ||