siRNA Details
| Gene Symbol: Rnf130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037658.1 | NP_001032747.1 | 791 | |||
| GenBank mRNA | AY190520.1 | - | 791 | |||
| BC166595.1 | - | 956 | 5599048 | |||
| FQ234228.1 | - | 281 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf130| Gene Symbol: Rnf130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037658.1 | NP_001032747.1 | 791 | |||
| GenBank mRNA | AY190520.1 | - | 791 | |||
| BC166595.1 | - | 956 | 5599048 | |||
| FQ234228.1 | - | 281 | ||||