siRNA Details
| Gene Symbol: Rnf130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037658.1 | NP_001032747.1 | 864 | |||
| GenBank mRNA | AY190520.1 | - | 864 | |||
| BC166595.1 | - | 1029 | 5599048 | |||
| FQ234228.1 | - | 2 | 354 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf130| Gene Symbol: Rnf130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037658.1 | NP_001032747.1 | 864 | |||
| GenBank mRNA | AY190520.1 | - | 864 | |||
| BC166595.1 | - | 1029 | 5599048 | |||
| FQ234228.1 | - | 2 | 354 | |||