siRNA Details
| Gene Symbol: Spag11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037850.1 | NP_001032939.1 | 2, 3 | 219 | ||
| NM_001037852.1 | NP_001032941.1 | 2, 3 | 279 | |||
| GenBank mRNA | DQ012092.1 | - | 2, 3 | 219 | ||
| DQ012093.1 | - | 2, 3 | 279 | |||
| Gene Symbol: Spag11bl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728419.4 | XP_002728465.2 | 2, 3 | 406 | ||
| XM_006253377.2 | XP_006253439.1 | 2, 3 | 409 | |||
| GenBank mRNA | AY600145.1 | - | 2, 3 | 239 | ||

Spag11b
Genomic Map