siRNA Details
| Gene Symbol: Zdhhc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039346.1 | NP_001034435.1 | 5, 6 | 837 | ||
| XM_006231403.2 | XP_006231465.1 | 5, 6 | 873 | |||
| XM_006231405.2 | XP_006231467.1 | 4, 5 | 679 | |||
| XM_006231407.2 | XP_006231469.1 | 4, 5 | 679 | |||
| GenBank mRNA | AY886532.1 | - | 5, 6 | 837 | ||
| BC127439.1 | - | 5, 6 | 838 | 8367792 | ||

Zdhhc16
Genomic Map