siRNA Details
Gene Symbol: Teddm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039915.2 | NP_001035004.1 | 1 | 244 | ||
GenBank mRNA | AJ515382.1 | - | 1 | 245 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Teddm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039915.2 | NP_001035004.1 | 1 | 244 | ||
GenBank mRNA | AJ515382.1 | - | 1 | 245 |