siRNA Details
| Gene Symbol: Igh-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC092580.1 | - | 711 | 7368318 | ||
| BC092586.1 | - | 2 | 686 | 7366480 | ||
| BC097958.1 | - | 2 | 667 | |||
| BC105825.1 | - | 3 | 680 | 7374168 | ||
| Gene Symbol: LOC314509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC092582.1 | - | 705 | |||
| Gene Symbol: LOC366747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC088184.1 | - | 5 | 667 | ||
| Gene Symbol: LOC678701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC158816.1 | - | 329 | |||

Igh-6
Genomic Map