siRNA Details
| Gene Symbol: Zc3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001053214.4 | XP_001053214.2 | 10 | 1397 | ||
| XM_006228482.1 | XP_006228544.1 | 10 | 1569 | |||
| XM_006228483.2 | XP_006228545.1 | 11 | 2106 | |||
| XM_006228484.2 | XP_006228546.1 | 11 | 1460 | |||
| XM_006228485.2 | XP_006228547.1 | 11 | 1789 | |||
| XM_006228486.2 | XP_006228548.1 | 10 | 1961 | |||
| XM_006228487.1 | XP_006228549.1 | 10 | 1320 | |||
| XM_006228488.2 | XP_006228550.1 | 11 | 1672 | |||
| XM_006228489.1 | XP_006228551.1 | 11 | 1459 | |||
| XM_006228491.2 | XP_006228553.1 | 11 | 1583 | |||
| XM_006228493.1 | XP_006228555.1 | 10 | 1569 | |||
| XM_008759031.1 | XP_008757253.1 | 10 | 1514 | |||

Zc3h4
Genomic Map